Accessories for microfiltration assemblies, ULTRA-WARE®, KIMBLE®
Leverandør: DWK Life Sciences
Accessories for microfiltration assemblies, ULTRA-WARE®, KIMBLE®
Leverandør: DWK Life Sciences
Certifikater
Gå til:
Om denne vare
9 tilgængelige muligheder nedenfor
Product Details & Documents
Product Family Options
Produktinformation
- TilgængelighedPrice
- SCERSP953753-0000Anodised aluminium clamp, 47 mm
- SCERSP953753-0090Anodised aluminium clamp, 90 mm
- SCERSP953752-0001Fritted glass support base, 47 mm
- SCERSP953826-0000Fritted glass support base, 47 mm, outer joint 40/35 NSFritted glass support base, 47 mm, outer joint 40/35 NSLog ind for tilgængelighed1 * 1 Items6.030,00 kr
- SCERSP953752-0090Fritted glass support base, 90 mm
- SCERSP953781-0090Glass funnel, graduated from 300 to 1000 ml in 50 ml intervals, 90 mm, 1000 mlGlass funnel, graduated from 300 to 1000 ml in 50 ml intervals, 90 mm, 1000 mlLog ind for tilgængelighed1 * 1 Items4.860,00 kr
- SCERSP971101-0347SPE funnel, 47 mm, 300 ml
- SCERSP971102-0047SPE support base, 47 mm
- SCERSP953906-0047Standard pickup adapter, 47 mm, with PTFE tubing of outer Ø 6,4 mmStandard pickup adapter, 47 mm, with PTFE tubing of outer Ø 6,4 mmLog ind for tilgængelighed1 * 1 Items6.080,00 kr