Orders LinkContactUsLinkComponent
Microinjectors, Nanoject III™
Microinjectors, Nanoject III™
Nanoject III™, 100 - 240 V for US, UK, Europe and Australia
Katalog # DRUM3-000-207
Leverandør:   DRUMMOND
Microinjectors, Nanoject III™
Katalog # DRUM3-000-207
Leverandør:  DRUMMOND
Microinjectors, Nanoject III™
Nanoject III™, 100 - 240 V for US, UK, Europe and Australia
Price
66.460,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The Nanoject III™ microinjector is designed to perform single manual injections and multiple injection cycle recipes. The auto homing feature eliminates the need to rehome the unit and reduce jamming of the plunger.
  • User-friendly programmable touchscreen operation
  • Accurate injection volumes down to 0,6 nanoliters
  • Ensures accurate and consistent injection volumes with hydraulic technology
  • O-rings not required to secure the micropipette
  • Lower injection volume capability
Leveringsinformation: Supplied with 4 interchangeable adapter plugs.
Specifikationer
  • Beskrivelse:
    Nanoject III™, 100 - 240 V for US, UK, Europe and Australia
  • Cat. No.:
    DRUM3-000-207
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement wire plunger
    Replacement wire plunger
    DRUM3-000-000-207
    Log ind for tilgængelighed
    1 * 1 Items - kr211.00
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement collet kit with chuck, green seal, wire plunger, and allan wrench
    Replacement collet kit with chuck, green seal, wire plunger, and allan wrench
    DRUM3-000-030-K
    Log ind for tilgængelighed
    1 * 1 Each - kr1,640.00
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Footswitch with remote capability for fill, empty and inject functions
    Footswitch with remote capability for fill, empty and inject functions
    DRUM3-000-032
    Log ind for tilgængelighed
    1 * 1 Items - kr5,710.00
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Silicon seal, green
    Silicon seal, green
    DRUM3-000-030-S
    Log ind for tilgængelighed
    1 * 3 Items - kr523.00