Om denne vare
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*5 TOC-skruelåg til nedbrydning påkrævet.
2 tilgængelige muligheder nedenfor
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*5 TOC-skruelåg til nedbrydning påkrævet.
Documents
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- 1.14878.0001
- 1.14879.0001
- M-Clarity quality level:
- MQ100
- MQ100
Specifikationer
Product Family Options
Produktinformation
- OmrådeKemisk metodeTil enhedPaknTilgængelighedPrice
- 1.14878.0001TOC-kuvettetest*5TOC-kuvettetest*55,0 - 80,0 mg/l TOCIndicatorProve, Nova 30/60, Multy25 TestsLog ind for tilgængelighed1 * 1 Each2.790,00 krSpecifikationer:
More
Specifications
Cat. No.1.14878.0001M-Clarity quality levelMQ100 - 1.14879.0001TOC-kuvettetest*5TOC-kuvettetest*550 - 800 mg/l TOCIndicatorProve, Nova 30/60, Multy25 TestsLog ind for tilgængelighed1 * 1 Each2.790,00 krSpecifikationer:
More
Specifications
Cat. No.1.14879.0001M-Clarity quality levelMQ100
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- TOC, skruelåg til brug i TermoreactorTOC, skruelåg til brug i Termoreactor1.73500.0001Log ind for tilgængelighed1 * 6 Items - 1.050,00 kr
Produktinformation
- Katalog #KoncentrationPaknTilgængelighedPrice
- TOC iflg. EN 1484 H3TOC iflg. EN 1484 H31.09017.01001000 (mg/l)100 mLLog ind for tilgængelighed1 * 100 mL - 1.610,00 kr