Orders LinkContactUsLinkComponent
Dissolved oxygen standard solution
Dissolved oxygen standard solution
Iodate-Iodide standard solution, 0.00125 N equivalent 10 mg/L DO, 500 mL
Katalog # HACH40149
Leverandør:   Hach
Dissolved oxygen standard solution
Katalog # HACH40149
Leverandør:  Hach
CAS nummer:  
Dissolved oxygen standard solution
Dissolved oxygen standard solution
Iodate-Iodide standard solution, 0.00125 N equivalent 10 mg/L DO, 500 mL
CAS nummer:  
Price
450,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Specifikationer
Specifikationer
  • Pakn:
    500 mL
  • Beskrivelse:
    Iodate-Iodide standard solution, 0.00125 N equivalent 10 mg/L DO