Tannin USP
Supplier: Spectrum Chemical
Jump to:
About this item
Synonyms: Gallotannin, Tannin
- CAS Number:1401-55-4
- Formula:C₇₆H₅₂O₄₆
- MDL Number:MFCD00066397
- Molecular Weight:1701.2 g/mol
- Merck Index:12,09221
- Melting Pt:218…219 °C
- Density:>1 g/cm³ (20 °C)
- Flash Pt:199 °C
- EINECS:215-753-2
3 Options Available Below
Product Details & Documents
Synonyms: Gallotannin, Tannin
- CAS Number:1401-55-4
- Formula:C₇₆H₅₂O₄₆
- MDL Number:MFCD00066397
- Molecular Weight:1701.2 g/mol
- Merck Index:12,09221
- Melting Pt:218…219 °C
- Density:>1 g/cm³ (20 °C)
- Flash Pt:199 °C
- EINECS:215-753-2