Pentylacetat
Leverandør: Spectrum Chemical
Om denne vare
Synonymer: n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate
- CAS nummer:628-63-7
- Formel:C₇H₁₄O₂
- MDL nummer:MFCD00009500
- Molecular Weight:130.19 g/mol
- Kogende Pt:149 °C (1013 hPa)
- Smeltende Pt:–71 °C
- Tæthed:0,876 g/cm³ (20 °C)
- Flash Pt:23 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:211-047-3
- FN:1104
- ADR:3,III
2 tilgængelige muligheder nedenfor
Product Details & Documents
Synonymer: n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate
- CAS nummer:628-63-7
- Formel:C₇H₁₄O₂
- MDL nummer:MFCD00009500
- Molecular Weight:130.19 g/mol
- Kogende Pt:149 °C (1013 hPa)
- Smeltende Pt:–71 °C
- Tæthed:0,876 g/cm³ (20 °C)
- Flash Pt:23 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:211-047-3
- FN:1104
- ADR:3,III
Specifications for Product Family
Specifikationstestresultater
- Assay:98% Min.