Supelco® Spectroquant® Test Kits, Copper Cell Test
Copper cell test, 0,05 - 8,00 mg/l Cu, Cuprizone, Prove, Nova 30/60, Move, 25 Tests
: Merck
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60 A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
For the determination of the total content of copper, please use the Crack Set (1.14688.0001) before the photometric procedure.
- Kemisk metode:Cuprizone
- Pakn:25 Tests
- Beskrivelse:Copper cell test
- Til enhed:Prove, Nova 30/60, Move
- Område:0,05 - 8,00 mg/l Cu
- Cat. No.:1.14553.0001
- M-Clarity quality level:MQ100