Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tazobactam ≥94%

Tazobactam ≥94%

1 g
Katalog # J66483.03
Irriterende
Leverandør:   Thermo Fisher Scientific
Tazobactam ≥94%
Katalog # J66483.03
CAS nummer:  89786-04-9
Tazobactam ≥94%

1 g

CAS nummer:  89786-04-9
Price
1.550,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

Purity: 94% CAS No.: 89786-04-9 Molecular Formula: C10H12N4O5S Formula Weight: 300.29 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White to off-white Melting Point: 115-145° Boiling Point: 77° Solubility: Soluble in water MDL No.: MFCD00867002

Applications: Increases efficacy of beta-lactam antibiotics by inactivating beta-lactamase enzymes of beta-lactam resistant microbes

Synonymer:  (2S,3S,5R)-3-((1H-1,2,3-Triazol-1-yl)methyl)-3-methyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide, Tazobactam acid
  • CAS nummer:89786-04-9
  • Formel:C₁₀H₁₂N₄O₅S
  • MDL nummer:MFCD00867002
  • Molecular Weight:300.289 g/mol
  • Kogende Pt:77 °C
  • Smeltende Pt:115…145 °C
  • Flash Pt:>110 °C (230 °F)
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakn:
    1 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.