Vials
Vial with 2, 5, 10, 15, 20 and 25 ml graduations marks, for LeadTrak™, Lead DR 100, FF3, IR24, BC3, SADT, 25,00 ml, Yes
Katalog # HACH219300
Leverandør: Hach
Gå til:
Product Details & Documents
Vial with 2, 5, 10, 15, 20 and 25 ml marks, for LeadTrak™, Lead DR 100, FF3, IR24, BC3, SADT.
Specifikationer
- Kasket inkluderet:Yes
- Kapacitet:25,00 ml
- Beskrivelse:Vial with 2, 5, 10, 15, 20 and 25 ml graduations marks, for LeadTrak™, Lead DR 100, FF3, IR24, BC3, SADT