BOD bottles, disposable
BOD bottle, 300 ml
Katalog # HACH2943100
Leverandør: Hach
Gå til:
Product Details & Documents
PET, clear.
- No cleaning required
- Unbreakable and recyclable
- Oxygen will not diffuse in or out of sample
These disposable bottles are fresh and ready to use.
Certifications: EPA compliant.
Bestillingsoplysninger: Stoppers are sold separately.
Specifikationer
- Kapacitet:300 ml
- Beskrivelse:BOD bottle
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Bottle stoppers, acrylic, unbreakable, for 300 ml BOD disposable bottlesBottle stoppers, acrylic, unbreakable, for 300 ml BOD disposable bottlesHACH2943900Log ind for tilgængelighed1 * 25 Items - 2.280,00 kr