Orders LinkContactUsLinkComponent
Electrode filling solution
Electrode filling solution
Internal filling solution for IntelliCAL™ PHC301 pH probes and MTC301 ORP probes, 3,0M KCl, saturated with silver chloride (AgCl), 28 ml bottle, 28 mL
Katalog # HACH2841700
Leverandør:   Hach
Electrode filling solution
Katalog # HACH2841700
Leverandør:  Hach
CAS nummer:  
Electrode filling solution
Electrode filling solution
Internal filling solution for IntelliCAL™ PHC301 pH probes and MTC301 ORP probes, 3,0M KCl, saturated with silver chloride (AgCl), 28 ml bottle, 28 mL
CAS nummer:  
Price
230,00 kr
Log ind for tilgængelighed
1 * 28 mL
Product Details & Documents
Specifikationer
Specifikationer
  • Pakn:
    28 mL
  • Beskrivelse:
    Internal filling solution for IntelliCAL™ PHC301 pH probes and MTC301 ORP probes, 3,0M KCl, saturated with silver chloride (AgCl), 28 ml bottle