Orders LinkContactUsLinkComponent
Hygiene cloths, Multipurpose Super
Hygiene cloths, Multipurpose Super
Leverandør:   PGI NONWOVEN
Hygiene cloths, Multipurpose Super
Leverandør:  PGI NONWOVEN
Certifikater
Hygiene cloths, Multipurpose Super
Hygiene cloths, Multipurpose Super
6 tilgængelige muligheder nedenfor
Product Details & Documents
These hardwearing towels are ideal for carrying out routine cleaning tasks in kitchens and restaurants. Because of their excellent absorbency the Multipurpose Super cloths are perfect for cleaning spills, both hot and cold.
  • Very low linting, ensuring no fibre shedding
  • High bulk and thickness, very absorbent
  • Heat resistant (210 °C): Suitable for carrying hot plates and pans when dry, can also be used for cleaning the stove
  • FCC approved: Completely safe when they come into contact with food, directly or indirectly
  • Machine washable up to 90 °C
Product Family Options
Produktinformation
  • Farve
    L×B
    Pakket
    Tilgængelighed
    Price
  • Perforated roll
    blå
    400×320 mm
    500 sheets/roll
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 2.960,00 kr
    2.960,00 kr
    1 * 1 ROL
       2.960,00 kr
  • M folded
    blå
    340×530 mm
    130 sheets/pack
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 130 Items
    1 * 130 Items - 940,00 kr
    940,00 kr
    1 * 130 Items
       940,00 kr
  • M folded
    hvid
    340×530 mm
    130 sheets/pack
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 130 Items
    1 * 130 Items - 899,00 kr
    899,00 kr
    1 * 130 Items
       899,00 kr
  • Perforated roll
    hvid
    400×320 mm
    500 sheets/roll
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 ROL
    1 * 1 ROL - 2.640,00 kr
    2.640,00 kr
    1 * 1 ROL
       2.640,00 kr
  • 1/4 folded
    blå
    340×320 mm
    260 sheets/pack
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 260 Items
    1 * 260 Items - 1.080,00 kr
    1.080,00 kr
    1 * 260 Items
       1.080,00 kr
  • 1/4 folded
    hvid
    340×320 mm
    260 sheets/pack
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 260 Items
    1 * 260 Items - 1.120,00 kr
    1.120,00 kr
    1 * 260 Items
       1.120,00 kr