Orders LinkContactUsLinkComponent
Reagents for comparators
Reagents for comparators
Supplier:   Palintest
Reagents for comparators
Supplier:  Palintest
Certificates
Reagents for comparators
Reagents for comparators
2 Options Available Below
Product Details & Documents
Comparator tablet reagents are designed for use with the contour comparator, standard comparator and other visual test platforms.
Product Family Options
Product Information
  • Beskrivelse
    Kemisk metode
    Pakn
    Availability
    Price
  • DPD 1 (free chlorine), 250 tests
    DPD
    250 Tab.
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 409,00 kr
    409,00 kr
    1 * 1 Each
       409,00 kr
  • pH (phenol red), 250 tests
    Phenol red
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 249,00 kr
    249,00 kr
    1 * 1 Each
       249,00 kr