Testkits, Guldtest<br /> til Spectroquant®, Supelco®
Katalog # 1.14821.0002
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Documents
Merck Segment Brochure
Produktlitteratur
msds_114821
SDS
Specifikationer
Specifikationer
- Kemisk metode:Rhodamine B
- Pakn:75 Tests
- Beskrivelse:Guldtest
- Til enhed:Prove, Nova 60
- Område:0,5 - 12,0 mg/l Au
- Cat. No.:1.14821.0002
- M-Clarity quality level:MQ100