Membrane filter funnels, Microfil
100 ml, 0,45 µm, 47 mm
Katalog # HACH2586300
Leverandør: Hach
Gå til:
Product Details & Documents
Mixed-cellulose ester, white, gridded.
- Sterile and ready-to-use
- Graduated
- Creates leak-proof seal when seated onto microfil stand alone support
Designed to eliminate cleaning and sanitisation of filter funnels between uses. Ideal for testing water samples from potable, non-potable, source, ambient, industrial, food, beverage, pharmaceutical and wastewater source.
Packaging: Individually packed.
Specifikationer
- Porestørrelse:0,45 µm
- Kapacitet:100 ml
- Til filter Ø:47 mm