Orders LinkContactUsLinkComponent
Volatile acid standards
Volatile acid standard solution, pack of 16×10 ml voluette ampoules, 62500 mg/l as acetic acid, 16
  705-2015
 :   Hach
Volatile acid standards
  705-2015
 :  Hach
 :  
Volatile acid standards
Volatile acid standards
Volatile acid standard solution, pack of 16×10 ml voluette ampoules, 62500 mg/l as acetic acid, 16
 :  
602,00 kr
1 * 16 Items
Acetate standard solution for accuracy checks in volatile acids determinations.
  • Koncentration:
    62500 mg/l as acetic acid
  • Pakn:
    16
  • Beskrivelse:
    Volatile acid standard solution, pack of 16×10 ml voluette ampoules