Orders LinkContactUsLinkComponent
Sulphite standard solution
Sulphite standard solution
Leverandør:   Hach
Sulphite standard solution
Leverandør:  Hach
Certifikater
Sulphite standard solution
Sulphite standard solution
2 tilgængelige muligheder nedenfor
Product Details & Documents
This equivalent standard is prepared with stabilised sodium thiosulfate, since sulphite is not a stable in solution.
Documents
Product Family Options
Produktinformation
  • Koncentration
    Pakn
    Tilgængelighed
    Price
  • HACH2408449
    Sulfite standard solution, 500 ml
    Sulfite standard solution, 500 ml
    15 mg/l as SO3
    500 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 598,00 kr
    598,00 kr
    1 * 500 mL
       598,00 kr
  • HACH2267410
    Sulfite standard solution, pack of 16×10 ml voluette ampoules (NIST)
    Sulfite standard solution, pack of 16×10 ml voluette ampoules (NIST)
    5000 mg/l as SO3
    16
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 16 Items
    1 * 16 Items - 602,00 kr
    602,00 kr
    1 * 16 Items
       602,00 kr