Orders LinkContactUsLinkComponent
Nickel standard solution
Nickel standard solution (NIST), 1000 mg/l as Ni, 100 mL
Katalog # HACH1417642
Leverandør:   Hach
Nickel standard solution
Katalog # HACH1417642
Leverandør:  Hach
CAS nummer:  
Nickel standard solution
Nickel standard solution
Nickel standard solution (NIST), 1000 mg/l as Ni, 100 mL
CAS nummer:  
Price
310,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
Standard solution for accuracy checks in nickel determinations.
Specifikationer
Specifikationer
  • Koncentration:
    1000 mg/l as Ni
  • Pakn:
    100 mL
  • Beskrivelse:
    Nickel standard solution (NIST)
Specifikationer