Handheld digital microscopes, special light, stroboscopic
About this item
Portable microscopes with stroboscopic technology to capture fast moving objects.
- Perfect pictures thanks to reduced motion blur, even under higher magnification
- Stroboscopic light mode can be switched on and off
- High speed USB 2.0 output
- Edge series model with high-quality optics provides a very sharp, bright and natural colour image with very low aberration and vignetting
- AM3715TB features exchangeable caps and an external trigger to synchronise picture capturing
- Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
Ideal for use in monitoring production lines in a manufacturing environment, or observing subjects such as a tadpole in a laboratory.
Delivery information: Supplied with carrying pouch and software (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)).
Caution: Please note: For optimal results the environmental lighting should be lower than normal and the microscope should be closer to the object being viewed.
2 Options Available Below
Product Details & Documents
Portable microscopes with stroboscopic technology to capture fast moving objects.
- Perfect pictures thanks to reduced motion blur, even under higher magnification
- Stroboscopic light mode can be switched on and off
- High speed USB 2.0 output
- Edge series model with high-quality optics provides a very sharp, bright and natural colour image with very low aberration and vignetting
- AM3715TB features exchangeable caps and an external trigger to synchronise picture capturing
- Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
Ideal for use in monitoring production lines in a manufacturing environment, or observing subjects such as a tadpole in a laboratory.
Delivery information: Supplied with carrying pouch and software (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)).
Caution: Please note: For optimal results the environmental lighting should be lower than normal and the microscope should be closer to the object being viewed.
Documents
Dino-Lite Powerful Portable Versatile Dino-Lite Software
Product Literature
Product Family Options
Product Information
- BeskrivelseModelForstørrelseBelysningOpløsningAvailabilityPrice
- Microscope with stroboscopic light, 60 fpsAM3713TB10 - 70×, 200×8 LEDsVGA (640×480)Log in for availability1 * 1 Itemskr3,610.00
- Microscope with stroboscopic light, Edge, 30 fpsAM3715TB20 - 200×8 LEDsVGA (640×480)Log in for availability1 * 1 Itemskr4,020.00
Accessories
Product Information
- Catalog #BeskrivelseB×D×HAvailabilityPrice
- IDCPRK-06ATabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EX150×220×270 mmLog in for availability1 * 1 Items - 1.880,00 kr
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- 630-2923Wi-Fi streamer with operating manual, power adapter and 1880 Ah Li-Ion batteryLog in for availability1 * 1 Items - 2.460,00 kr
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- 630-1821Foot pedal, enables users to take a picture with their footLog in for availability1 * 1 Items - 472,00 kr
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- 630-2517Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal armLog in for availability1 * 1 Items - 2.540,00 kr