Cell spreaders, L-shaped
Celleudstrygningsspatler, L-formede, engangs, sterile, 146 mm, 10 stk./pose
Catalog # HEATHS8171B
Supplier: HEATHROW
Jump to:
Product Details & Documents
ABS disposable L-shaped cell spreader with smooth surface that will not tear agar plate or disturb media layer. Supplied sterile for immediate use without flaming.
Specifications
- Pakket:10 stk./pose
- Beskrivelse:Celleudstrygningsspatler, L-formede, engangs, sterile
- Længde:146 mm