Orders LinkContactUsLinkComponent
Needles, single use, PrecisionGlide™
Needles, single use, PrecisionGlide™
Katalog # BDAM305165
Leverandør:   BD Medical
Needles, single use, PrecisionGlide™
Katalog # BDAM305165
Leverandør:  BD Medical
Needles, single use, PrecisionGlide™
Needles, single use, PrecisionGlide™
Price
2.500,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
BD PrecisionGlide™ needle technology incorporates a thin wall, tri-bevel tip and a proprietary lubricant which is designed to provide ease of penetration.