Inverted microscope, CKX53
Inverted microscope, Trinocular
Katalog # OLYMCKX53TR-PH
Leverandør: OLYMPUS OPTICAL
Gå til:
Product Details & Documents
Ergonomically designed and enhanced imaging for cell culture, the CKX53 is designed for cell sampling and manipulation, especially for fluorescence observations.
- Observations in lightfield transmission and phase contrast evolutionary to fluorescence
- High contrast thanks to integrated phase system (IPC) (without ring change)
- Three-position fluorescence illuminator: Two sets of filters for excitation (blue and green), empty position, manual shutter and neutral filter slide 6%
Compatible with CKX3-SLP and IX2-SL condenser slides, CKX3-SPLIC, CKX3-SLPAS, IX2-SPLC, IX2-SLPH1, IX2-SLPH2 optical elements, CKX3-MVR object guide, CKX3-HO35DM Petri dish rack, reflective fluorescence illuminator and mercury, metal halide or LED light sources.
: Included CACHN 10× / 0,25 Ph / IPC lens, power cable, platinum insert, Allen key, power supply, two camera cable clips with screws - contrast-inverted 3-D views, optional.
Specifikationer
- Model:Trinocular
- Beskrivelse:Inverted microscope
- Kontrastteknik:Brightfield, phase contrast and fluorescence
- Cat. No.:OLYMCKX53TR-PH
- Revolver:4 positioner
- Hoved:45° vinklet
- Kondensor:0,30
- Belysning:LED
- Arbejdsafstand:72 – 194 mm
- Fokuseringssystem:Koaksial grov og finindstilling
- Krydsbord:200×252 mm
- Okularer:WF 10×/22 mm
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Dust Cover for CKX Microscopes
Produktinformation
- Katalog #TilgængelighedPrice
- CK2-SS stage extension plate for CK, CKX and IX microscopesCK2-SS stage extension plate for CK, CKX and IX microscopesOLYM030362Log ind for tilgængelighed1 * 1 Items - 554,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Power cable with elbow
Produktinformation
- Katalog #TilgængelighedPrice
- CKX53SF microscope frame (base body)CKX53SF microscope frame (base body)OLYMN5228200Log ind for tilgængelighed1 * 1 Items - 34.840,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- CKX3-HOUN, Universal holder (slides and dishes) for CKX3-MVRCKX3-HOUN, Universal holder (slides and dishes) for CKX3-MVROLYMN5229000Log ind for tilgængelighed1 * 1 Items - 4.190,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- CKX3-SLP phase contrast slider, pre-centered for CKX53 and IX53CKX3-SLP phase contrast slider, pre-centered for CKX53 and IX53OLYMN5228500Log ind for tilgængelighed1 * 1 Items - 716,00 kr