Orders LinkContactUsLinkComponent
VWR®, Microscope Cleaning Set
VWR®, Microscope Cleaning Set
Cleaning kit (for models SZBL360, SZTL360, SZB360 OH, SZT360 OH, SZBL360-Plus, SZTL360-Plus, SZB360, SZT360)
Katalog # 630-2081
Leverandør:   VWR Collection
VWR®, Microscope Cleaning Set
Katalog # 630-2081
Leverandør:  VWR Collection
VWR®, Microscope Cleaning Set
VWR®, Microscope Cleaning Set
Cleaning kit (for models SZBL360, SZTL360, SZB360 OH, SZT360 OH, SZBL360-Plus, SZTL360-Plus, SZB360, SZT360)
Price
240,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This cleaning set includes:
- Lens liquid
- Cotton sticks (×5)
- Anti-static cloth (18×15 cm)
- Smart wipes (×2)
- Air pump
Documents
Specifikationer
  • Beskrivelse:
    Cleaning kit (for models SZBL360, SZTL360, SZB360 OH, SZT360 OH, SZBL360-Plus, SZTL360-Plus, SZB360, SZT360)
  • Seq:
    999