Latex gloves, Rotiprotect®
Marketsource
Leverandør: Roth Carl
Om denne vare
These disposable gloves are suitable for protection against complex risks and for food processing applications.
- Powder-free, smooth finish
- Very smooth and pleasantly soft
- Extremely stretchy
- High tactile sensitivity
- Thiuram-free
Certifications: EN 374; Cat. III, AQL 1,5
Food contact approved
2 tilgængelige muligheder nedenfor
Product Details & Documents
These disposable gloves are suitable for protection against complex risks and for food processing applications.
- Powder-free, smooth finish
- Very smooth and pleasantly soft
- Extremely stretchy
- High tactile sensitivity
- Thiuram-free
Certifications: EN 374; Cat. III, AQL 1,5
Food contact approved
Specifications for Product Family
Specifikationer
- Catalog No:
- 112-0040
- 112-0042
- Ambidextrous/Hånd-specifik:
- Ambidextrous
- Ambidextrous
- Engangs/Genbrugelig:
- Engangs
- Engangs
- Latex/latex-fri:
- Latex
- Latex
- Med pudder/pudderfri:
- Pudderfri
- Pudderfri
- Overflade:
- Smooth
- Smooth
- Manchet:
- Rullet kant
- Rullet kant
- EN norm:
- EN 374
- EN 374
- Godkendt til fødevarekontakt:
- +
- +
Specifikationer
Product Family Options
Produktinformation
- Handske størrelseFarveLængdeTykkelsePakketTilgængelighedPrice
- Specifikationer:Marketsource
More
Specifications
Cat. No.112-0040Ambidextrous/Hånd-specifikAmbidextrousEngangs/GenbrugeligEngangsLatex/latex-friLatexMed pudder/pudderfriPudderfriOverfladeSmoothManchetRullet kantEN normEN 374Godkendt til fødevarekontakt+Marketsource - Specifikationer:Marketsource
More
Specifications
Cat. No.112-0042Ambidextrous/Hånd-specifikAmbidextrousEngangs/GenbrugeligEngangsLatex/latex-friLatexMed pudder/pudderfriPudderfriOverfladeSmoothManchetRullet kantEN normEN 374Godkendt til fødevarekontakt+Marketsource