Orders LinkContactUsLinkComponent
Retention time standards
Leaking underground storage tank retention time standard includes n-hexane (C6), n-octacosane (C28), n-decane (C10), n-triacontane (C30), n-dodecane (C12), n-tetracontane (C40), n-tetracosane (C24), with 25 µg/ml each in methylene chloride, 1 ml/ampule, 1
Katalog # REST31200
Leverandør:   Restek
Retention time standards
Katalog # REST31200
Leverandør:  Restek
CAS nummer:  
Retention time standards
Leaking underground storage tank retention time standard includes n-hexane (C6), n-octacosane (C28), n-decane (C10), n-triacontane (C30), n-dodecane (C12), n-tetracontane (C40), n-tetracosane (C24), with 25 µg/ml each in methylene chloride, 1 ml/ampule, 1
CAS nummer:  
Price
388,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Used during initial sample screening to determine retention time windows for each petroleum product.
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    Leaking underground storage tank retention time standard includes n-hexane (C6), n-octacosane (C28), n-decane (C10), n-triacontane (C30), n-dodecane (C12), n-tetracontane (C40), n-tetracosane (C24), with 25 µg/ml each in methylene chloride, 1 ml/ampule
Specifikationer