Orders LinkContactUsLinkComponent
Packing table
Packing table
Packing table, mobil, 1447 ×750× 900 mm
Catalog # HUPF7500184
Supplier:   HUPFER METALLWERKE
Packing table
Catalog # HUPF7500184
Supplier:  HUPFER METALLWERKE
Packing table
Packing table
Packing table, mobil, 1447 ×750× 900 mm
Price
12.180,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
This packing table is made up of high-quality stainless steel (both top and bottom). It can also be also as a work bench to pack surgical instruments.
Specifications
  • Beskrivelse:
    Packing table, mobil
  • B×D×H:
    1447 ×750× 900 mm
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • HUPF7500185
    Hook rail, double, stainless steel, 1250×40×1447 mm, single-side for packing table
    Log in for availability
    1 * 1 Items - 4.020,00 kr