Orders LinkContactUsLinkComponent
Immersion heater
Immersion heater
Immersion heater for use with 230 V mains supply
Catalog # BINH1003566
Marketsource
Supplier:   3B Scientific
Immersion heater
Catalog # BINH1003566
Supplier:  3B Scientific
Marketsource
Immersion heater
Immersion heater
Immersion heater for use with 230 V mains supply
Price
114,29 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Immersion heater with overheating protection (VDE-compliant). Important: operates solely on a mains voltage of 230 V.
Specifications
  • Beskrivelse:
    Immersion heater for use with 230 V mains supply