Anti-FEZF2 Rabbit Polyclonal Antibody
100 μg
Katalog # BOSSBS-12148R
Leverandør: Bioss
Gå til:
Product Details & Documents
Transcription repressor. Required for the specification of corticospinal motor neurons and other subcerebral projection neurons. May play a role in layer and neuronal subtype-specific patterning of subcortical projections and axonal fasciculation. Controls the development of dendritic arborization and spines of large layer V pyramidal neurons. May be involved in innate immunity (By similarity).
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ZNF312
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Documents
bs-12148R
Datablad
Specifikationer
- Størrelse:100 μg
- Western blot:Yes
- Clonality:Polyclonal
- Gen ID:55079
- Purification:Purified by Protein A.
- Konjugation:Unconjugated
- Form:liquid
- Isotype:IgG
- Antigen synonymer:ZFP312|ZNF312|TOF|FEZL|FKSG36|FEZ
- Koncentration:1 ug/ul
- Shipping temperature:4 °C
- Antigen name:FEZ family zinc finger 2
- Cat. No.:BOSSBS-12148R
- Reactivity:Human|Mouse
- Immunogen:1-50/459
- Vært:Rabbit
- ImmunoChemistry:Yes
- Antistof type:Primary
- Antigen symbol:FEZF2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.



