Cork Borer Sets
Cork borers, set of 26, 5,0 - 60,0 mm
Katalog # JUCHVL02079
Leverandør: JUCHHEIM Laborgerate GmbH
Gå til:
Product Details & Documents
These cork borers are made of brass and have solid handles.
Specifikationer
- Hul Ø:5,0 - 60,0 mm
- Beskrivelse:Cork borers, set of 26
- Seq:07