Shoe covers, CPE
blå, U*
HOIT110176
These disposable shoe covers are made of CPE.
- Soft and durable fabric
- Latex-free
- Embossed
- With elasticated opening
- Lightweight, breathable, dustproof and waterproof
Ideal for use in hospitals, food processing, manufacturing, construction and for hygienic applications.
* One size fits all.
: Packed in a box.
- Skostørrelse:U*
- Farve:blå
- Cat. No.:HOIT110176
- Outfit applikationer:Laboratory|Food