Glass marker
Glass marker, length 135 mm, weight 20 g
Katalog # JUCHVL01837
Leverandør: JUCHHEIM Laborgerate GmbH
Gå til:
Product Details & Documents
Glass marker provided along with diamond and case.
Specifikationer
- Beskrivelse:Glass marker, length 135 mm, weight 20 g