Calibration weight sets, class E2, Proof Line
These class E2 weight sets include aluminium (1 to 5 mg) or nickel silver (10 to 500 mg) or stainless steel (1 g and higher) weights.
These class E2 weight sets include aluminium (1 to 5 mg) or nickel silver (10 to 500 mg) or stainless steel (1 g and higher) weights.
Product Family Options
E2 Weight sets without certificate
- VægtsætModel
- 1 mg - 500 mgWithout certificate1 * 1 Itemskr4,050.00
- 1 g - 200 gWithout certificate1 * 1 Itemskr10,280.00
- 1 g - 1 kgWithout certificate1 * 1 Itemskr16,440.00
- 1 g - 5 kgWithout certificate1 * 1 Itemskr32,500.00
E2 Weight sets with DAkkS certificate
- VægtsætModel
- 1 mg - 500 mgWith DAkkS certificate1 * 1 Itemskr7,290.00
- 1 g - 200 gWith DAkkS certificate1 * 1 Itemskr13,250.00
- 1 g - 1 kgWith DAkkS certificate1 * 1 Itemskr19,790.00
- 1 g - 5 kgWith DAkkS certificate1 * 1 Itemskr37,370.00