SPE flow filters, Resprep®
Katalog # REST26024
Leverandør: Restek
Specifikationer
- Beskrivelse:Flow filters
- Pakn:20
Specifikationer
Om denne vare
Flow filters are designed to improve flow when filtering oil and grease samples. These filters can be used with Resprep® Diskcover-47 reservoir or any 47 mm glass sample reservoir.