Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Septa
Septa for screw thread vials, Target®
Septa for screw thread vials, Target®
Septa for screw thread vials, Target®
Catalog # NSCAC4010-60
Supplier:
THERMO NATIONAL SCIENTIFIC
Expand image
Septa for screw thread vials, Target®
Catalog # NSCAC4010-60
Supplier:
THERMO NATIONAL SCIENTIFIC
Specifications
Ø:
10 mm
Septum:
Red PTFE/white silicone, pre-slit
Beskrivelse:
Septa, for 10-425 screw thread caps
Seq:
05
Pakn:
100
Specifications
About this item
Product Details
Product Details