Orders LinkContactUsLinkComponent
X-Gal
X-Gal
Leverandør:   G-Biosciences
X-Gal
Leverandør:  G-Biosciences
Certifikater
3 tilgængelige muligheder nedenfor
Product Details & Documents
5-Bromo-4-chloro-3-indolyl beta-D-galactopyranoside is most commonly known as X-Gal. X-gal is a histochemical substrate for beta-galactosidase which cleaves X-Gal to yield an insoluble blue precipitate.
  • Molecular Biology Grade
  • Purity: Greater than 99% (HPLC)
  • Solution: Clear (2% in DMF)
  • Biological Activity: Blue colour with Lambda, pUC, and M13
X-Gal is a mainstay in molecular biology cloning applications in which it is used to detect the activity of beta-galactosidase. X-Gal is used to detect the insertion of foreign DNA into the lacZ region of a plasmid DNA. Insertion of DNA into the lacZ region results in the loss of beta-galactosidase activity. Bacteria cells that retain active beta-galactosidase will result in characteristic blue colonies. Successful disruption of the lacZα gene disrupts the α-complemtation of the beta-galactosidase gene and the precipitate does not form, resulting in white colonies.
Many other applications also use X-Gal as a substrate to detect beta-galactosidase activity. These include beta-galactosidase -antibody linked immunoassays and immunohistochemistry, coliphage detection based on beta-galactosidase induction and the detection of micrometastasis formation during tumor progession.
X-gal is commonly used in conjunction with IPTG for blue/white screening.
Synonymer:  BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
  • CAS nummer:7240-90-6
  • Formel:C₁₄H₁₅BrClNO₆
  • MDL nummer:MFCD00005666
  • Molecular Weight:408,63 g/mol
Product Family Options
Produktinformation
  • Pakning
    Pakn
    Tilgængelighed
    Price
  • Vial
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mg
    1 * 100 mg - 412,00 kr
    412,00 kr
    1 * 100 mg
       412,00 kr
  • Vial
    1 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 g
    1 * 1 g - 721,00 kr
    721,00 kr
    1 * 1 g
       721,00 kr
  • Bottle
    10 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 g
    1 * 10 g - 4.010,00 kr
    4.010,00 kr
    1 * 10 g
       4.010,00 kr