Shakers, purging method
TOC shaker, TOC-X5, for purging method
Katalog # HACHLQV1489900001
Leverandør: Hach
Gå til:
Product Details & Documents
Shaker which is used for purging of inorganic carbon (TIC). It has an automatic shutoff system. Accommodates eight cuvettes simultaneously.
Specifikationer
- Beskrivelse:TOC shaker, TOC-X5, for purging method
- Cat. No.:HACHLQV1489900001
- B×D×H:190×240×135 mm
- Vægt:2,9 kg
- Model:TOC-X5
- Rystebevægelse:Rocking/Rotating