PCR plates, 96-well, without skirt, pre-loaded with PCR strips of 8 tubes, Multiply®
Catalog # SARS72.985
Supplier: SARSTEDT
Specifications
- Ant. brønde:96
- Profil:Standard
- Skørt:Uden skørt
- Brøndfarve:klar
- Beskrivelse:PCR plate, pre-loaded with PCR strips of 8 tubes (0,2 ml)
- Pakn:20