Orders LinkContactUsLinkComponent
NBB®-C, Concentrated Broth
NBB®-C, Concentrated Broth
NBB®-concentrate (NBB®-C), 250 ml, -, 250 ml, Bottle, 9
Katalog # 710-0851
Leverandør:   DOEHLER GROUP
NBB®-C, Concentrated Broth
Katalog # 710-0851
Leverandør:  DOEHLER GROUP
CAS nummer:  
NBB®-C, Concentrated Broth
NBB®-C, Concentrated Broth
NBB®-concentrate (NBB®-C), 250 ml, -, 250 ml, Bottle, 9
CAS nummer:  
Price
1.460,00 kr
Log ind for tilgængelighed
1 * 9 Items
Product Details & Documents
Concentrated, selective liquid medium for fast trace detection in unfiltered beer. Under its NBB® brand, Döhler offers innovative culture media for the fast and reliable detection of beverage spoiling microorganisms, such as Lactobacilli, Pectinatus and Megasphaera, in all samples that arise in breweries, as well as for the wine industry. NBB®-C is a liquid culture medium with enhanced selectivity through the addition of yeast-cloudy beer samples.
  • Simply added to the beer sample for analysis
  • Final microscopic inspection, no indicator verification
Specifikationer
Specifikationer
  • Pakning:
    Bottle
  • Pakn:
    9
  • Standard:
    -
  • Beskrivelse:
    NBB®-concentrate (NBB®-C), 250 ml
  • Påfyldning:
    250 ml