Orders LinkContactUsLinkComponent
NBB®-B, Broth
NBB®-B, Broth
NBB®-bouillon (NBB®-B), -, 250 ml, alle prøvetyper i bryggeriindustrien, Bottle, 9
Catalog # 710-0741
Supplier:   DOEHLER GROUP
NBB®-B, Broth
Catalog # 710-0741
Supplier:  DOEHLER GROUP
CAS Number:  
NBB®-B, Broth
NBB®-B, Broth
NBB®-bouillon (NBB®-B), -, 250 ml, alle prøvetyper i bryggeriindustrien, Bottle, 9
CAS Number:  
Price
1.340,00 kr
Log in for availability
1 * 9 Items
Product Details & Documents
Selective liquid medium for fast trace detection in yeast culture. Under its NBB® brand, Döhler offers innovative culture media for the fast and reliable detection of beverage spoiling microorganisms, such as Lactobacilli, Pectinatus and Megasphaera, in all samples that arise in breweries, as well as for the wine industry. NBB®-B is a liquid culture media with indicator dye for detection of beer spoiling microorganisms in yeast samples or as an enrichment medium.
  • Direct use of liquid medium
  • Simple, visual screening by an indicator change from red to yellow
Documents
Specifications
Specifications
  • Pakning:
    Bottle
  • Pakn:
    9
  • Standard:
    -
  • Beskrivelse:
    NBB®-bouillon (NBB®-B)
  • Påfyldning:
    250 ml
  • Til:
    alle prøvetyper i bryggeriindustrien