Normal foot
Normal foot, 0,4 kg, 130×240×90 mm
Katalog # BINH1000354
Marketsource
Leverandør: 3B Scientific
Gå til:
Product Details & Documents
Superficial structures of the human foot are shown in this high quality model.
The internal bones, muscles, ligaments and nerves are also represented in this normal foot skeleton.
Specifikationer
- Vægt:0,4 kg
- Beskrivelse:Normal foot
- B×D×H:130×240×90 mm