Frocks
hvid, XL
Katalog # MAKA4574702
Leverandør: MUTEXIL
Gå til:
Product Details & Documents
Disposable non woven frocks with collar. Suitable for use in industrial and food environments.
- With zip closure
- Without pockets
Weight: 40 g/m²
Specifikationer
- Tøj størrelse:XL
- Farve:hvid
- Seq:01
- Køn:Unisex
- Cat. No.:MAKA4574702
- Vægt:40 g/m²
- Outfit applikationer:Laboratory|Food
- Lukkestørrelse:Med lynlås
- Lommer:uden lommer