Horizontal wafer shippers, 150 mm SmartStack®
Material: STAT-PRO® 100M, sort
Katalog # ENTEHWS15010161C2M
Leverandør: ENTEGRIS
Gå til:
Product Details & Documents
Horizontal wafer shipper.
- Packaged, double bagged, heat sealed
- Automation compatible
Specifikationer
- Farve:sort
- Beskrivelse:Material: STAT-PRO® 100M