VWR®, Cryoboxes
Divider for boxes, fiberboard, 16-cell, 4×4
Catalog # 478-0241
Supplier: VWR Collection
Jump to:
Product Details & Documents
Polycarbonate.
- Can be used from –190 to +121 °C
VWR® freezer boxes can be used with most manufacturer’s upright and chest freezer racks in general purpose and ultra-low mechanical freezers.
Documents
VWR for Cell Culture
Product Literature
Specifications
- Rum:4×4
- Beskrivelse:Divider for boxes, fiberboard, 16-cell