Orders LinkContactUsLinkComponent
Agarose, SeaPrep™
Agarose, SeaPrep™
Agarose, SeaPrep™
Catalog # LONZ50302
Supplier:   LONZA
Agarose, SeaPrep™
Catalog # LONZ50302
Supplier:  LONZA
Agarose, SeaPrep™
Agarose, SeaPrep™
Price
5.920,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Ultra-soft agarose, ideal for high efficiency hybridoma cloning and expansion of cDNA libraries.
Specifikationer
  • Beskrivelse:
    Agarose, SeaPrep™
Accessories
Produktinformation
  • Catalog #
    Beskrivelse
    Availability
    Price
  • LONZ50302
    Agarose, SeaPrep™
    Log ind for tilgængelighed
    1 * 25 g - 5.920,00 kr