Orders LinkContactUsLinkComponent
Accessories for adhesion test kits
Leverandør:   Industrial Physics
Accessories for adhesion test kits
Leverandør:  Industrial Physics
Certifikater
Accessories for adhesion test kits
Accessories for adhesion test kits
11 tilgængelige muligheder nedenfor
Product Details & Documents
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • SIMESP1702
    Spare cutting blade, 6 teeths, 1 mm, acc. to ISO 2409, ASTM D3359, suitable for CC2000 and CC3000 test kits
    Spare cutting blade, 6 teeths, 1 mm, acc. to ISO 2409, ASTM D3359, suitable for CC2000 and CC3000 test kits
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 984,00 kr
    984,00 kr
    1 * 1 Items
       984,00 kr
  • SIMESP1705
    Spare cutting blade, 11 teeths, 1 mm, acc. to ISO 2409, suitable for CC2000 and CC3000 test kits
    Spare cutting blade, 11 teeths, 1 mm, acc. to ISO 2409, suitable for CC2000 and CC3000 test kits
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 984,00 kr
    984,00 kr
    1 * 1 Items
       984,00 kr
  • SIMESP1706
    Spare cutting blade, 11 teeths, 1,5 mm, acc. to ASTM D3359 <2009, suitable for CC2000 and CC3000 test kits
    Spare cutting blade, 11 teeths, 1,5 mm, acc. to ASTM D3359 <2009, suitable for CC2000 and CC3000 test kits
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 984,00 kr
    984,00 kr
    1 * 1 Items
       984,00 kr
  • SIMESP1703
    Spare cutting blade, 6 teeths, 2 mm, acc. to ISO 2409, ASTM D3359, suitable for CC2000 and CC3000 test kits
    Spare cutting blade, 6 teeths, 2 mm, acc. to ISO 2409, ASTM D3359, suitable for CC2000 and CC3000 test kits
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 984,00 kr
    984,00 kr
    1 * 1 Items
       984,00 kr
  • SIMESP1704
    Spare cutting blade, 6 teeths, 3 mm, acc. to ISO 2409, suitable for CC2000 and CC3000 test kits
    Spare cutting blade, 6 teeths, 3 mm, acc. to ISO 2409, suitable for CC2000 and CC3000 test kits
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 984,00 kr
    984,00 kr
    1 * 1 Items
       984,00 kr
  • SIMEVF2355
    Spare knife, round, 1 mm, acc. to ISO 2409 & ASTM D3359, suitable for C1000 test kits
    Spare knife, round, 1 mm, acc. to ISO 2409 & ASTM D3359, suitable for C1000 test kits
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.590,00 kr
    2.590,00 kr
    1 * 1 Items
       2.590,00 kr
  • SIMEVF2359
    Spare knife, round, 1 mm, acc. to ASTM D3359 <2009, suitable for C1000 test kits
    Spare knife, round, 1 mm, acc. to ASTM D3359 <2009, suitable for C1000 test kits
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.830,00 kr
    2.830,00 kr
    1 * 1 Items
       2.830,00 kr
  • SIMEVF1861
    Spare knife, round, 1,5 mm acc. to ASTM D3359 <2009, suitable for C1000 test kits
    Spare knife, round, 1,5 mm acc. to ASTM D3359 <2009, suitable for C1000 test kits
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.950,00 kr
    2.950,00 kr
    1 * 1 Items
       2.950,00 kr
  • SIMEVF2357
    Spare knife, round, 2 mm, acc. to ISO 2409 & ASTM D3359, suitable for C1000 test kits
    Spare knife, round, 2 mm, acc. to ISO 2409 & ASTM D3359, suitable for C1000 test kits
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.590,00 kr
    2.590,00 kr
    1 * 1 Items
       2.590,00 kr
  • SIMEVF2358
    Spare knife, round, 3 mm, acc. to ISO 2409, suitable for C1000 test kits
    Spare knife, round, 3 mm, acc. to ISO 2409, suitable for C1000 test kits
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.590,00 kr
    2.590,00 kr
    1 * 1 Items
       2.590,00 kr
  • SIMESP1710
    Nylon brush for cross cut adhesion test
    Nylon brush for cross cut adhesion test
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 138,00 kr
    138,00 kr
    1 * 1 Items
       138,00 kr
Accessories
Produktinformation
  • Katalog #
    Bredde
    Tilgængelighed
    Price
  • Tape for cross cut adhesion test, 3 rolls of 11,0 m, acc. to ISO 2409:2003
    Tape for cross cut adhesion test, 3 rolls of 11,0 m, acc. to ISO 2409:2003
    SIMESP3010
    25,0 mm
    Log ind for tilgængelighed
    1 * 3 ROL - 285,00 kr
Produktinformation
  • Katalog #
    Bredde
    Tilgængelighed
    Price
  • Tape for cross cut adhesion test, 1 roll of 11,0 m, acc. to ISO 2409:2003
    Tape for cross cut adhesion test, 1 roll of 11,0 m, acc. to ISO 2409:2003
    SIMESP3007
    25,0 mm
    Log ind for tilgængelighed
    1 * 1 Items - 193,00 kr