Drug testing kit for surfaces
Multipanel 5 drug test for surfaces (AMP/COC/MAMP/MDMA/OPI) with swab/buffer
Katalog # SUREPLAB4DS24300_U
Leverandør: SURESCREEN
Gå til:
Product Details & Documents
Using the swab and buffer, this kit allows swabbing of suspect surfaces or powders to determine if they are amphetamines.
- Testing drug paraphernalia, surfaces or suspected raw drug
- Highly accurate and easy to use
- Results within five minutes
Suitable for customs, security, or anywhere where drug use is a concern.
Specifikationer
- Beskrivelse:Multipanel 5 drug test for surfaces (AMP/COC/MAMP/MDMA/OPI) with swab/buffer
- Seq:04