Syringe for vapour pressure osmometer
KNAUA0433
: KNAUER
- Volumen:1000 µl
- Beskrivelse:Syringe with stainless steel piston
- Pakn:1
This syringe has a glass barrel, stainless steel piston with PTFE seal, including needle with spring stopper.
This syringe has a glass barrel, stainless steel piston with PTFE seal, including needle with spring stopper.