Orders LinkContactUsLinkComponent
Quality control test suspensions
Quality control test suspensions
 :   CROSSTEX
Quality control test suspensions
 :  CROSSTEX
Quality control test suspensions
The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.
  • 10 ml pro vial
  • 100 dose
Documents
Product Family Options
  • Beskrivelse
    Pakn
  • Bacillus subtilis
    10 mL
    1 * 10 mL
    1 * 10 mL - 2.830,00 kr
    2.830,00 kr
    1 * 10 mL
       2.830,00 kr
  • Clostridium sporogenes
    10 mL
    1 * 10 mL
    1 * 10 mL - 2.780,00 kr
    2.780,00 kr
    1 * 10 mL
       2.780,00 kr
  • Geobacillus stearothermophilus
    10 mL
    1 * 10 mL
    1 * 10 mL - 4.520,00 kr
    4.520,00 kr
    1 * 10 mL
       4.520,00 kr
  • Aspergillus brasiliensis
    10 mL
    1 * 10 mL
    1 * 10 mL - 4.740,00 kr
    4.740,00 kr
    1 * 10 mL
       4.740,00 kr
  • Bacillus atrophaeus
    10 mL
    1 * 10 mL
    1 * 10 mL - 4.610,00 kr
    4.610,00 kr
    1 * 10 mL
       4.610,00 kr