Membrane disc filters
GN-6, grid, individually packed, without pad, 0,45 µm, 47 mm
Katalog # HACH1353001
Leverandør: Hach
Gå til:
Product Details & Documents
These GN-metrical membrane filters are ideal for testing recreational water, wastewater and drinking water. They meets U.S. Environmental Protection Agency (USEPA) and American Society for Testing and Materials (ASTM) specifications to ensure complete retention and maximum recovery of bacteria.
- Convenient and pre-sterilised
- Gridded design
Specifikationer
- Ø:47 mm
- Porestørrelse:0,45 µm
- Model:GN-6, grid, individually packed, without pad
- Cat. No.:HACH1353001