Orders LinkContactUsLinkComponent
Sink table
Sink table
Laboratory sink table, with drain valve 38,1 mm, 500×600×660 mm
Katalog # BOCH21800
Leverandør:   BOCHEM
Sink table
Katalog # BOCH21800
Leverandør:  BOCHEM
Sink table
Sink table
Laboratory sink table, with drain valve 38,1 mm, 500×600×660 mm
Price
20.090,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This laboratory sink table is made of stainless steel 18/10.
Specifikationer
  • Beskrivelse:
    Laboratory sink table, with drain valve 38,1 mm
  • B×D×H:
    500×600×660 mm